4-Chloro-N,N-diisopropylbenzamide
Supplier: Apollo Scientific
|
Warning
|
OR59402-1G
OR59402-25G
OR59402-5G
APOSOR59402-1GEA
72.7
EUR
APOSOR59402-1G
APOSOR59402-25G
APOSOR59402-5G
4-Chloro-N,N-diisopropylbenzamide
4-Chloro-N,N-diisopropylbenzamide
Formula:
C₁₃H₁₈ClNO MW: 239.75 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00032340 CAS Number: 79606-45-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...