1-(Phenylsulphonyl)pyrrolidine
Supplier: Apollo Scientific
Warning
OR59413-1G OR59413-25G OR59413-5G
APOSOR59413-1GEA 72.7 EUR
APOSOR59413-1G APOSOR59413-25G APOSOR59413-5G
1-(Phenylsulphonyl)pyrrolidine
1-(Phenylsulphonyl)pyrrolidine
Formula: C₁₀H₁₃NO₂S
Boiling Pt: 92…96 °C/0.04mmHg
Melting Pt: 154…156 °C
Density: 1.2077 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD01055979
CAS Number: 5033-22-7