N-Isopropylhydroxylamine hydrochloride 98%
Supplier: Apollo Scientific
Warning
OR59437-5G OR59437-100G OR59437-25G
APOSOR59437-5GEA 118 EUR
APOSOR59437-5G APOSOR59437-100G APOSOR59437-25G
N-Isopropylhydroxylamine hydrochloride 98%
N-Isopropylhydroxylamine hydrochloride
Formula: C₃H₉NO
MW: 111.57 g/mol
Melting Pt: 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00012599
CAS Number: 50632-53-6