m-Anisic acid 98+%

Supplier: Apollo Scientific
Warning

APOSOR5944-500GEA 98.9 EUR
APOSOR5944-500G
m-Anisic acid 98+%
m-Anisic acid
Formula: C₈H₈O₃
MW: 152,15 g/mol
Boiling Pt: 170…172 °C/10mmHg
Melting Pt: 105…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00002499
EINECS: 209-574-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR