Methyl-2-(3-aminophenyl)acetate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 2-(3-aminophenyl)acetate
OR59446-25G OR59446-5G
APOSOR59446-25GEA 517 EUR
APOSOR59446-25G APOSOR59446-5G
Methyl-2-(3-aminophenyl)acetate
Methyl-2-(3-aminophenyl)acetate
Formula: C₉H₁₁NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD07366754
CAS Number: 52913-11-8