4-Nitro-o-toluic acid
Supplier: Apollo Scientific
Warning
OR59451-5G OR59451-25G OR59451-100G
APOSOR59451-5GEA 21.9 EUR
APOSOR59451-5G APOSOR59451-25G APOSOR59451-100G
4-Nitro-o-toluic acid
4-Nitro-o-toluic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Melting Pt: 150…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00210697
CAS Number: 1975-51-5
EINECS: 217-828-5