2,4-Dibromopyrimidine 95%
Supplier: Apollo Scientific
Warning
OR59452-10G OR59452-5G OR59452-1G
APOSOR59452-10GEA 288 EUR
APOSOR59452-10G APOSOR59452-5G APOSOR59452-1G
2,4-Dibromopyrimidine 95%
2,4-Dibromopyrimidine
Formula: C₄H₂Br₂N₂
MW: 237.88 g/mol
Melting Pt: 65…67 °C
Storage Temperature: Ambient
MDL Number: MFCD06798232
CAS Number: 3921-01-5