1,4-Dibromo-2,5-diiodobenzene 95%
Supplier: Apollo Scientific
Warning
OR59457-1G OR59457-25G OR59457-5G
APOSOR59457-1GEA 66.9 EUR
APOSOR59457-1G APOSOR59457-25G APOSOR59457-5G
1,4-Dibromo-2,5-diiodobenzene 95%
1,4-Dibromo-2,5-diiodobenzene
Formula: C₆H₂Br₂I₂
MW: 487.6970 g/mol
Melting Pt: 167 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00082898
CAS Number: 63262-06-6