Methyl-2-furoate
Supplier: Apollo Scientific
Danger
Synonyms: Methyl 2-Furancarboxylate, Methyl furan-2-carboxylate, Methyl 2-furoate
OR5946-100G OR5946-25G OR5946-500G
APOSOR5946-100GEA 92.8 EUR
APOSOR5946-100G APOSOR5946-25G APOSOR5946-500G
Methyl-2-furoate
Methyl 2-furoate
Formula: C₆H₆O₃
Boiling Pt: 181 °C
Density: 1.179 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00003236
CAS Number: 611-13-2
EINECS: 210-254-6
UN: 2810
ADR: 6.1,III