Methyl-5-chloropyrazine-2-carboxylate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 5-chloropyrazine-2-carboxylate
OR59463-100G OR59463-25G OR59463-5G
APOSOR59463-100GEA 364 EUR
APOSOR59463-100G APOSOR59463-25G APOSOR59463-5G
Methyl-5-chloropyrazine-2-carboxylate 98%
Methyl 5-chloropyrazine-2-carboxylate
Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
Boiling Pt: 76…78 °C/1mmHg
Melting Pt: 90.5…91.5 °C
Storage Temperature: Ambient
MDL Number: MFCD01632102
CAS Number: 33332-25-1