3-Amino-N-benzylbenzenesulphonamide 97%
Supplier: Apollo Scientific
Warning
OR59784-1G OR59784-5G
APOSOR59784-1GEA 94.5 EUR
APOSOR59784-1G APOSOR59784-5G
3-Amino-N-benzylbenzenesulphonamide 97%
3-Amino-N-benzylbenzenesulphonamide
Formula: C₁₃H₁₄N₂O₂S
Melting Pt: 72…73 °C
Storage Temperature: Ambient
MDL Number: MFCD07363817
CAS Number: 303780-52-1