Diethylene glycol 99%
Supplier: Apollo Scientific
Warning
OR59808-10KG OR59808-2.5KG
APOSOR59808-10KGEA 329 EUR
APOSOR59808-10KG APOSOR59808-2.5KG
Diethylene glycol 99%
Diethylene glycol
Formula: C₄H₁₀O₃
Boiling Pt: 245…246 °C
Melting Pt: –10 °C
Density: 1.118 g/cm³ (25 °C)
Flash Pt: 123 °C
Storage Temperature: Ambient
MDL Number: MFCD00002882
CAS Number: 111-46-6
EINECS: 203-872-2