3-Chlorobenzonitrile 99%
Supplier: Apollo Scientific
Warning
Synonyms: m-Chlorobenzonitrile
OR5983-100G OR5983-25G
APOSOR5983-100GEA 132 EUR
APOSOR5983-100G APOSOR5983-25G
3-Chlorobenzonitrile 99%
3-Chlorobenzonitrile
Formula: C₇H₄ClN
MW: 137.57 g/mol
Boiling Pt: 94 °C/10mmHg
Melting Pt: 39…41 °C
Storage Temperature: Ambient
MDL Number: MFCD00001798
CAS Number: 766-84-7