3-Formylbenzoic acid 95%
Supplier: Apollo Scientific
Warning
Synonyms: Isophthalaldehydic acid, 3-Carboxybenzaldehyde
OR59888-25G OR59888-100G
APOSOR59888-25GEA 150 EUR
APOSOR59888-25G APOSOR59888-100G
3-Formylbenzoic acid 95%
3-Formylbenzoic acid
Formula: C₈H₆O₃
Melting Pt: 173…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00039575
CAS Number: 619-21-6
EINECS: 210-585-6