4-Bromo-2,6-xylenol 95+%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-2,6-dimethylphenol, 5-Bromo-2-hydroxy-3-xylene
OR59890-100G OR59890-25G OR59890-500G
APOSOR59890-100GEA 46.4 EUR
APOSOR59890-100G APOSOR59890-25G APOSOR59890-500G
4-Bromo-2,6-xylenol 95+%
4-Bromo-2,6-xylenol
Formula: C₈H₉BrO
MW: 201.06 g/mol
Melting Pt: 74…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00002314
CAS Number: 2374-05-2
EINECS: 219-153-1