1-(p-Toluenesulphonyl)imidazole
Supplier: Apollo Scientific
Warning
OR59904-5G OR59904-100G OR59904-25G OR59904-500G
APOSOR59904-5GEA 14.7 EUR
APOSOR59904-5G APOSOR59904-100G APOSOR59904-25G APOSOR59904-500G
1-(p-Toluenesulphonyl)imidazole
1-(p-Toluenesulphonyl)imidazole
Formula: C₁₀H₁₀N₂O₂S
MW: 222.26 g/mol
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00005285
CAS Number: 2232-08-8