1,2,4,5-Tetramethyl-3,6-dinitrobenzene 97+%
Supplier: Apollo Scientific
Warning
Synonyms: 1,4-Dinitrodurene, Dinitrodurene
OR59914-25G OR59914-5G OR59914-1G
APOSOR59914-25GEA 195 EUR
APOSOR59914-25G APOSOR59914-5G APOSOR59914-1G
1,2,4,5-Tetramethyl-3,6-dinitrobenzene 97+%
1,2,4,5-Tetramethyl-3,6-dinitrobenzene
Formula: C₁₀H₁₂N₂O₄
MW: 224.22 g/mol
Melting Pt: 208…211 °C
Storage Temperature: Ambient
MDL Number: MFCD00007164
CAS Number: 5465-13-4
EINECS: 226-766-8