2-Bromo-5-methoxyaniline 97%
Supplier: Apollo Scientific
Warning
OR59916-100G OR59916-25G OR59916-5G
APOSOR59916-100GEA 115 EUR
APOSOR59916-100G APOSOR59916-25G APOSOR59916-5G
2-Bromo-5-methoxyaniline 97%
2-Bromo-5-methoxyaniline
Formula: C₇H₈BrNO
MW: 202.0500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070741
CAS Number: 59557-92-5