Neocuproine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2,9-Dimethyl-1,10-phenanthroline
OR59924-100G OR59924-25G OR59924-5G
APOSOR59924-100GEA 420 EUR
APOSOR59924-100G APOSOR59924-25G APOSOR59924-5G
Neocuproine 98%
Neocuproine
Formula: C₁₄H₁₂N₂
MW: 208.26 g/mol
Melting Pt: 159 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004973
CAS Number: 484-11-7
EINECS: 207-601-9