2,2,6,6-Tetramethylpiperidinooxy (TEMPO) free radical
Supplier: Apollo Scientific
Danger
Synonyms: TEMPO, 2,2,6,6-Tetramethyl-1-oxylpiperidine, 2,2,6,6-Tetramethylpiperidine 1-oxyl
OR59926-100G OR59926-25G OR59926-500G
APOSOR59926-100GEA 46.4 EUR
APOSOR59926-100G APOSOR59926-25G APOSOR59926-500G
2,2,6,6-Tetramethylpiperidinooxy (TEMPO) free radical
2,2,6,6-Tetramethylpiperidinooxy (TEMPO, free radical)
Formula: C₉H₁₈NO
MW: 156.25 g/mol
Melting Pt: 36…40 °C
Flash Pt: 67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009599
CAS Number: 2564-83-2
EINECS: 219-888-8
UN: 3263
ADR: 8 II