5-Bromo-1H-pyrrolo[2,3-b]pyridine 98%
Supplier: Apollo Scientific
Danger
Synonyms: 5-Bromo-7-azaindole
OR59927-25G OR59927-100G
APOSOR59927-25GEA 53.8 EUR
APOSOR59927-25G APOSOR59927-100G
5-Bromo-1H-pyrrolo[2,3-b]pyridine 98%
5-Bromo-1H-pyrrolo[2,3-b]pyridine
Formula: C₇H₅BrN₂
MW: 197.04 g/mol
Melting Pt: 178…179 °C
Storage Temperature: Ambient
MDL Number: MFCD06659677
CAS Number: 183208-35-7