1-[4-(2-Methyl-1H-imidazol-1-yl)phenyl]methanamine 95%
Supplier: Apollo Scientific
Synonyms: 4-(2-Methyl-1H-imidazol-1-yl)benzylamine
OR59932-1G OR59932-5G OR59932-250MG
APOSOR59932-1GEA 435 EUR
APOSOR59932-1G APOSOR59932-5G APOSOR59932-250MG
1-[4-(2-Methyl-1H-imidazol-1-yl)phenyl]methanamine 95%
1-[4-(2-Methyl-1H-imidazol-1-yl)phenyl]methanamine
Formula: C₁₁H₁₃N₃
Melting Pt: 90.5…91.5 °C
Storage Temperature: Ambient
MDL Number: MFCD06617944
CAS Number: 883291-45-0