5-Hydroxy-3,4-dimethoxybenzaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 3-Hydroxy-4,5-dimethoxybenzaldehyde, 3,4-Dimethoxy-5-hydroxybenzaldehyde
OR59937-5G OR59937-10G
APOSOR59937-5GEA 116 EUR
APOSOR59937-5G APOSOR59937-10G
5-Hydroxy-3,4-dimethoxybenzaldehyde
5-Hydroxy-3,4-dimethoxybenzaldehyde
Formula: C₉H₁₀O₄
MW: 182.1733 g/mol
Boiling Pt: 177…180 °C/12mmHg
Melting Pt: 67…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00010442
CAS Number: 29865-90-5