3-Acetyl-2-methyl-1,6-naphthyridine 95+%
Supplier: Apollo Scientific
Warning
OR5996-1G OR5996-10G OR5996-5G
APOSOR5996-1GEA 273 EUR
APOSOR5996-1G APOSOR5996-10G APOSOR5996-5G
3-Acetyl-2-methyl-1,6-naphthyridine 95+%
3-Acetyl-2-methyl-1,6-naphthyridine
Formula: C₁₁H₁₀N₂O
Melting Pt: 106…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00202914
CAS Number: 52816-65-6