1,1,1,3-Tetrachloropropane 98%
Supplier: Apollo Scientific
Warning
Synonyms: HC-250fb
OR59960-100G OR59960-25G
APOSOR59960-100GEA 65.4 EUR
APOSOR59960-100G APOSOR59960-25G
1,1,1,3-Tetrachloropropane 98%
1,1,1,3-Tetrachloropropane
Formula: Cl₃CCH₂CH₂Cl
MW: 181.88 g/mol
Boiling Pt: 159 °C/760mmHg
Melting Pt: –35 °C
Density: 1.47 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00054655
CAS Number: 1070-78-6
EINECS: 213-981-7
UN: 1993
ADR: 3,III