Glyoxal dimethyl acetal 60% in aqueous solution
Supplier: Apollo Scientific
Warning
Synonyms: Dimethoxyacetaldehyde, 2,2-Dimethoxyacetaldehyde, Dimethoxyethanal
OR59985-100G OR59985-500G OR59985-5G
APOSOR59985-100GEA 79.9 EUR
APOSOR59985-100G APOSOR59985-500G APOSOR59985-5G
Glyoxal dimethyl acetal 60% in aqueous solution
Glyoxal dimethyl acetal
Formula: OHCCH(OCH₃)₂
MW: 104.11 g/mol
Boiling Pt: 100…102 °C/759mmHg
Density: 1.15 g/cm³ (25 °C)
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00134410
CAS Number: 51673-84-8