2-Chloro-3-nitrotoluene 95%
Supplier: Apollo Scientific
Warning
OR59989-1G OR59989-5G OR59989-10G OR59989-25G
APOSOR59989-1GEA 14.7 EUR
APOSOR59989-1G APOSOR59989-5G APOSOR59989-10G APOSOR59989-25G
2-Chloro-3-nitrotoluene 95%
2-Chloro-3-nitrotoluene
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 147 °C/25mmHg
Melting Pt: 19…21 °C
Density: 1.298 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00134653
CAS Number: 3970-40-9
UN: 2433
ADR: 6.1 III