1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester
Supplier: Apollo Scientific
Warning
Synonyms: 1-(Tetrahydropyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 1-(Tetrahydro-2H-pyran-2-yl)-1H-pyrazole-5-boronic acid Pinacol ester, 1-(Tetrahydropyran-2-yl)-1H-pyrazole-5-boronic acid pinacol ester, 1-(Tetrahydro-2H-pyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
OR60006-100G
APOSOR60006-100GEA 75.4 EUR
APOSOR60006-100G
1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester
1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester
Formula: C₁₄H₂₃BN₂O₃
MW: 278.16 g/mol
Melting Pt: 74…78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09037501
CAS Number: 903550-26-5