7-Amino-2-methylindazole
Supplier: Apollo Scientific
Warning
OR60014-1G OR60014-5G OR60014-250MG
APOSOR60014-1GEA 24.8 EUR
APOSOR60014-1G APOSOR60014-5G APOSOR60014-250MG
7-Amino-2-methylindazole
7-Amino-2-methylindazole
Formula: C₈H₉N₃
MW: 147.18 g/mol
Melting Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD03787880
CAS Number: 90223-02-2