2-Acetyl-5-methylfuran
Supplier: Apollo Scientific
Warning
OR6003-100G OR6003-500G
APOSOR6003-100GEA 129 EUR
APOSOR6003-100G APOSOR6003-500G
2-Acetyl-5-methylfuran
2-Acetyl-5-methylfuran
Formula: C₇H₈O₂
Boiling Pt: 100…101 °C/25mmHg
Density: 1.066 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003243
CAS Number: 1193-79-9
EINECS: 214-779-1