4-Methyl-2-(methylthio)pyrimidine 97%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Methyl-2(methylsulphanyl)pyrimidine
OR60031-25G OR60031-5G
APOSOR60031-25GEA 219 EUR
APOSOR60031-25G APOSOR60031-5G
4-Methyl-2-(methylthio)pyrimidine 97%
4-Methyl-2-(methylthio)pyrimidine
Formula: C₆H₈N₂S
MW: 140.21 g/mol
Density: 1.007 g/cm³ (25 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00023242
CAS Number: 14001-63-9