3-Chloro-6-iodopyridazine
Supplier: Apollo Scientific
Warning
OR60058-1G OR60058-100G
APOSOR60058-1GEA 0 EUR
APOSOR60058-1G APOSOR60058-100G
3-Chloro-6-iodopyridazine
3-Chloro-6-iodopyridazine
Formula: C₄H₂ClIN₂
MW: 240.43 g/mol
Melting Pt: 110…120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08275187
CAS Number: 135034-10-5