2-Chloro-4-hydroxypyridine
Supplier: Apollo Scientific
Warning
OR60066-25G OR60066-5G OR60066-1G
APOSOR60066-25GEA 58 EUR
APOSOR60066-25G APOSOR60066-5G APOSOR60066-1G
2-Chloro-4-hydroxypyridine
2-Chloro-4-hydroxypyridine
Formula: C₅H₄ClNO
MW: 129.55 g/mol
Melting Pt: 169…172 °C
Storage Temperature: Ambient
MDL Number: MFCD01646108
CAS Number: 17368-12-6
EINECS: 241-399-3