2-Formyl-3-picoline 96%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Methylpyridine-2-carboxaldehyde, 3-Methylpicolinaldehyde
OR60091-1G OR60091-5G
APOSOR60091-1GEA 21.9 EUR
APOSOR60091-1G APOSOR60091-5G
2-Formyl-3-picoline 96%
2-Formyl-3-picoline
Formula: C₇H₇NO
Boiling Pt: 84 °C/12.8mmHg
Melting Pt: 8 °C
Density: 1.08 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD02181144
CAS Number: 55589-47-4