1H-Indazole-5-boronic acid
Supplier: Apollo Scientific
Warning
OR60111-1G OR60111-250MG
APOSOR60111-1GEA 56.7 EUR
APOSOR60111-1G APOSOR60111-250MG
1H-Indazole-5-boronic acid
1H-Indazole-5-boronic acid
Formula: C₇H₇BN₂O₂
MW: 161.9537 g/mol
Melting Pt: 175…180 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06739151
CAS Number: 338454-14-1