N-Methyl-4-(pyrrolidin-1-ylmethyl)benzylamine
Supplier: Apollo Scientific
Danger
OR60123-1G OR60123-250MG OR60123-500MG OR60123-2.5G
APOSOR60123-1GEA 510 EUR
APOSOR60123-1G APOSOR60123-250MG APOSOR60123-500MG APOSOR60123-2.5G
N-Methyl-4-(pyrrolidin-1-ylmethyl)benzylamine
N-Methyl-4-(pyrrolidin-1-ylmethyl)benzylamine
Formula: C₁₃H₂₀N₂
MW: 204.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08060619
CAS Number: 884507-40-8
UN: 2735
ADR: 8 III