1-Hydroxy-2-naphthoic acid
Supplier: Apollo Scientific
Danger
Synonyms: 1-Hydroxy-naphthalene-2-carboxylic acid
OR60129-100G OR60129-250G OR60129-5G OR60129-500G
APOSOR60129-100GEA 24.8 EUR
APOSOR60129-100G APOSOR60129-250G APOSOR60129-5G APOSOR60129-500G
1-Hydroxy-2-naphthoic acid
1-Hydroxy-2-naphthoic acid
Formula: C₁₁H₈O₃
MW: 188.18 g/mol
Boiling Pt: 375
Melting Pt: 195…200 °C (dec.)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00003960
CAS Number: 86-48-6
EINECS: 201-674-0
Merck Index: 13,04859