5-Bromo-4-methylpyridine-2-carboxylic acid 98%
Supplier: Apollo Scientific
|
Warning
|
OR60140-1G
OR60140-250MG
OR60140-5G
APOSOR60140-1GEA
119
EUR
APOSOR60140-1G
APOSOR60140-250MG
APOSOR60140-5G
5-Bromo-4-methylpyridine-2-carboxylic acid 98%
5-Bromo-4-methylpyridine-2-carboxylic acid
Formula:
C₇H₆BrNO₂ MW: 216.0300 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD07375100 CAS Number: 886365-02-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...