Ethyl-2-methyl-1,3-oxazole-4-carboxylate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 2-methyloxazole-4-carboxylate
OR60168-5G
APOSOR60168-5GEA 197 EUR
APOSOR60168-5G
Ethyl-2-methyl-1,3-oxazole-4-carboxylate 98%
Ethyl-2-methyl-1,3-oxazole-4-carboxylate
Formula: C₇H₉NO₃
MW: 155.15 g/mol
Boiling Pt: 210 °C/760mmHg
Melting Pt: 24…25 °C
Storage Temperature: Ambient
MDL Number: MFCD07367959
CAS Number: 10200-43-8