trans-4-Decenal
Supplier: Apollo Scientific
Warning
Synonyms: (E)-4-Decenal
OR6017-5G OR6017-1G
APOSOR6017-5GEA 50.9 EUR
APOSOR6017-5G APOSOR6017-1G
trans-4-Decenal
trans-4-Decenal
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 80 °C/4mmHg
Density: 0.842 g/cm³ (25 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00065447
CAS Number: 65405-70-1
EINECS: 265-741-6