2-Ethyl-4-methyl-1,3-dioxolane
Supplier: Apollo Scientific
Warning
OR60175-1G OR60175-250MG OR60175-500MG OR60175-5G
APOSOR60175-1GEA 141 EUR
APOSOR60175-1G APOSOR60175-250MG APOSOR60175-500MG APOSOR60175-5G
2-Ethyl-4-methyl-1,3-dioxolane
Formula: C₆H₁₂O₂
Boiling Pt: 117 °C
Flash Pt: 29 °C
Storage Temperature: Ambient
MDL Number: MFCD16622267
CAS Number: 4359-46-0
EINECS: 224-435-2
UN: 1993
ADR: 3,III