Pyrrole-2-carbonitrile 98%
Supplier: Apollo Scientific
Danger
OR60193-25G OR60193-100G
APOSOR60193-25GEA 72.5 EUR
APOSOR60193-25G APOSOR60193-100G
Pyrrole-2-carbonitrile 98%
Pyrrole-2-carbonitrile
Formula: C₅H₄N₂
MW: 92.101 g/mol
Boiling Pt: 92…94 °C/2mmHg
Density: 1.086 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Freezer
MDL Number: MFCD00234061
CAS Number: 4513-94-4
UN: 3276
ADR: 6.1 III