2,3-Dimethylpyrazine 99%
Supplier: Apollo Scientific
Warning
OR6020-10G OR6020-250G OR6020-50G
APOSOR6020-10GEA 46.6 EUR
APOSOR6020-10G APOSOR6020-250G APOSOR6020-50G
2,3-Dimethylpyrazine 99%
2,3-Dimethylpyrazine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 156 °C
Melting Pt: –12 °C
Density: 1.022 g/cm³ (25 °C)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00006144
CAS Number: 5910-89-4
EINECS: 227-630-0
UN: 1993
ADR: 3,III