tert-Butyl-2-(aminomethyl)pyrrolidine-1-carboxylate 98%
Supplier: Apollo Scientific
Danger
Synonyms: tert-Butyl 2-(aminomethyl)pyrrolidine-1-carboxylate, 2-(Aminomethyl)pyrrolidine, N1-BOC protected
OR60204-10G OR60204-25G OR60204-5G
APOSOR60204-10GEA 183 EUR
APOSOR60204-10G APOSOR60204-25G APOSOR60204-5G
tert-Butyl-2-(aminomethyl)pyrrolidine-1-carboxylate 98%
tert-Butyl-2-(aminomethyl)pyrrolidine-1-carboxylate
Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Density: 1.014 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01317793
CAS Number: 177911-87-4
UN: 2735
ADR: 8 III