Oxetan-3-ol 95%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Hydroxyoxetane
OR60206-1G OR60206-5G OR60206-25G
APOSOR60206-1GEA 21.9 EUR
APOSOR60206-1G APOSOR60206-5G APOSOR60206-25G
Oxetan-3-ol 95%
Oxetan-3-ol
Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 67…70 °C/8mmHg
Density: 1.125 g/cm³ (25 °C)
Flash Pt: 87.2 °C
Storage Temperature: Ambient
MDL Number: MFCD09056790
CAS Number: 7748-36-9