2,4-Dichlorothiazole 99%
Supplier: Apollo Scientific
Warning
OR60252-25G OR60252-100G OR60252-5G
APOSOR60252-25GEA 34.8 EUR
APOSOR60252-25G APOSOR60252-100G APOSOR60252-5G
2,4-Dichlorothiazole 99%
2,4-Dichlorothiazole
Formula: C₃HCl₂NS
MW: 154.0200 g/mol
Boiling Pt: 184 °C
Melting Pt: 44 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08691358
CAS Number: 4175-76-2