N1,N1-Dimethylbenzene-1,2-diamine
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-N,N-dimethylaniline
OR60254-1G OR60254-25G OR60254-5G
APOSOR60254-1GEA 79.9 EUR
APOSOR60254-1G APOSOR60254-25G APOSOR60254-5G
N1,N1-Dimethylbenzene-1,2-diamine
N1,N1-Dimethylbenzene-1,2-diamine
Formula: C₈H₁₂N₂
MW: 136.1943 g/mol
Boiling Pt: 223 °C
Density: 1.013 g/cm³ (25 °C)
Storage Temperature: Freezer
MDL Number: MFCD01706706
CAS Number: 2836-03-5