1-(2-Bromoethoxy)-2,4-dichlorobenzene
Supplier: Apollo Scientific
Warning
Synonyms: beta-Bromo-2,4-dichlorophenetole
OR6066-10G OR6066-1G OR6066-5G
APOSOR6066-10GEA 288 EUR
APOSOR6066-10G APOSOR6066-1G APOSOR6066-5G
1-(2-Bromoethoxy)-2,4-dichlorobenzene
1-(2-Bromoethoxy)-2,4-dichlorobenzene
Formula: C₈H₇BrCl₂O
MW: 269.95 g/mol
Boiling Pt: 120…130 °C/0.4mmHg
Storage Temperature: Ambient
MDL Number: MFCD00043464
CAS Number: 6954-77-4