8-Chlorotetrazolo[1,5-a]pyridine 98%
Supplier: Apollo Scientific
Warning
OR6102-100MG OR6102-250MG
APOSOR6102-100MGEA 43.5 EUR
APOSOR6102-100MG APOSOR6102-250MG
8-Chlorotetrazolo[1,5-a]pyridine 98%
8-Chlorotetrazolo[1,5-a]pyridine
Formula: C₅H₃ClN₄
Melting Pt: 194…200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00490111
CAS Number: 40971-88-8