Dimethyl sulphoxide 99.7+%
Supplier: Apollo Scientific
Warning
Synonyms: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO
OR61029-100ML OR61029-25ML OR61029-50ML
APOSOR61029-100MLEA 451 EUR
APOSOR61029-100ML APOSOR61029-25ML APOSOR61029-50ML
Dimethyl sulphoxide 99.7+%
Dimethyl sulphoxide
Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189 °C
Melting Pt: 16…19 °C
Density: 1.1 g/cm³ (25 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
Merck Index: 13,03285